Gene Transcripts

Gene Symbol
Rgs20
Entrez Gene ID
362477
Gene Name
regulator of G-protein signaling 20
Gene Aliases
Chromosome Location
Chr. 5: 14415841 - 14493744 on Build Rnor_6.0
UniGene ID
Rn.125947
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127495.1 NP_001120967.1 4 947
GenBank mRNA BC160842.1 - 4 947

View Full Product Details