Gene Transcripts

Gene Symbol
Dnajc25
Entrez Gene ID
362526
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 25
Gene Aliases
Chromosome Location
Chr. 5: 76150840 - 76171424 on Build Rnor_6.0
UniGene ID
Rn.82298
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025021.1 NP_001020192.1 3 856
GenBank mRNA BC091296.1 - 3 856

View Full Product Details