Gene Transcripts

Gene Symbol
Ccdc23
Entrez Gene ID
362578
Gene Name
coiled-coil domain containing 23
Gene Aliases
RGD1311232
Chromosome Location
Chr. 5: 138245639 - 138251353 on Build Rnor_6.0
UniGene ID
Rn.2954
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038992.1 NP_001034081.1 2 196
NM_001038994.2 NP_001034083.1 2 296
GenBank mRNA BC099226.1 - 2 196
BF559487.1 - 2 302

View Full Product Details