Gene Transcripts

Gene Symbol
Wdr43
Entrez Gene ID
362703
Gene Name
WD repeat domain 43
Gene Aliases
Chromosome Location
Chr. 6: 23433532 - 23473177 on Build Rnor_6.0
UniGene ID
Rn.101883
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037791.2 NP_001032880.2 2 299
XM_006239770.2 XP_006239832.1 2 307
GenBank mRNA BC169117.1 - 2 280

View Full Product Details