Gene Transcripts

Gene Symbol
Nfam1
Entrez Gene ID
362966
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
RGD1308316
Chromosome Location
Chr. 7: 123856718 - 123893792 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006242140.2 XP_006242202.1 2 358
XM_006242142.2 XP_006242204.1 2 271

View Full Product Details