Gene Transcripts

Gene Symbol
Ppp2r3a
Entrez Gene ID
363122
Gene Name
protein phosphatase 2, regulatory subunit B'', alpha
Gene Aliases
Chromosome Location
Chr. 8: 109481352 - 109621460 on Build Rnor_6.0
UniGene ID
Rn.93024
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012202.2 NP_001012202.2 11 3414
XM_006243633.2 XP_006243695.1 12 3716
XM_006243634.2 XP_006243696.1 11 2140
GenBank mRNA BC087097.1 - 11 1489

View Full Product Details