Gene Transcripts

Gene Symbol
Gemin8
Entrez Gene ID
363462
Gene Name
gem (nuclear organelle) associated protein 8
Gene Aliases
Fam51a1
Chromosome Location
Chr. X: 29806605 - 29825439 on Build Rnor_6.0
UniGene ID
Rn.18700
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007756.1 NP_001007757.1 4 648
GenBank mRNA BC079160.1 - 4 648

View Full Product Details