Gene Transcripts

Gene Symbol
Sgcz
Entrez Gene ID
364605
Gene Name
sarcoglycan zeta
Gene Aliases
Chromosome Location
Chr. 16: 57136433 - 58207615 on Build Rnor_6.0
UniGene ID
Rn.231585
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108875.2 NP_001102345.2 7 947
XM_008771302.1 XP_008769524.1 7 781

View Full Product Details