Gene Transcripts

Gene Symbol
MAP7
Entrez Gene ID
9053
Gene Name
microtubule associated protein 7
Gene Aliases
E-MAP-115, EMAP115
Chromosome Location
Chr. 6: 136342281 - 136551015 on Build GRCh38
UniGene ID
Hs.486548
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198608.1 NP_001185537.1 17 2424
NM_001198609.1 NP_001185538.1 17 2448
NM_001198611.1 NP_001185540.1 16 2313
NM_001198614.1 NP_001185543.1 18 2427
NM_001198615.1 NP_001185544.1 17 2157
NM_001198616.1 NP_001185545.1 16 2409
NM_001198617.1 NP_001185546.1 15 2238
NM_001198618.1 NP_001185547.1 17 2287
NM_001198619.1 NP_001185548.1 17 2160
NM_003980.4 NP_003971.1 17 2520
XM_006715598.2 XP_006715661.1 18 2969
XM_006715599.2 XP_006715662.1 18 2738
XM_006715600.2 XP_006715663.1 18 2714
XM_006715601.3 XP_006715664.1 17 2740
XM_011536243.2 XP_011534545.1 17 3339
XM_011536244.2 XP_011534546.1 16 3248
XM_011536245.1 XP_011534547.1 16 2127
XM_011536246.2 XP_011534548.1 16 3229
XM_017011471.1 XP_016866960.1 17 2834
GenBank mRNA AJ242501.1 - 15 1827
AJ242502.1 - 16 1998
AJ420541.1 - 5 401
AK294461.1 - 16 2083
AK295749.1 - 17 2176
AK296483.1 - 17 2287
AK296556.1 - 17 2286
AK299355.1 - 17 2157
AK312789.1 - 17 2120
AK316062.1 - 17 2160
AK316529.1 - 18 2293
BC025777.1 - 17 2116 5222987
X73882.1 - 17 2209

View Full Product Details