Gene Transcripts

Gene Symbol
Fastkd5
Entrez Gene ID
366192
Gene Name
FAST kinase domains 5
Gene Aliases
Fastkd5-ps1, RGD1563839
Chromosome Location
Chr. 3: 123154263 - 123170540 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762238.1 XP_008760460.1 2 1938

View Full Product Details