Gene Transcripts

Gene Symbol
Oxct2a
Entrez Gene ID
366463
Gene Name
3-oxoacid CoA transferase 2A
Gene Aliases
Chromosome Location
Chr. 5: 140813957 - 140815711 on Build Rnor_6.0
UniGene ID
Rn.110620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012221.1 NP_001012221.1 1 1547
GenBank mRNA BC083681.1 - 1 1547

Gene Symbol
LOC100361036
Entrez Gene ID
100361036
Gene Name
rCG31267-like
Gene Aliases
Chromosome Location
Chr. 5: 141016544 - 141018848 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764112.1 XP_008762334.1 1 2097

View Full Product Details