Gene Transcripts

Gene Symbol
Scrt1
Entrez Gene ID
366951
Gene Name
scratch family zinc finger 1
Gene Aliases
Chromosome Location
Chr. 7: 117583453 - 117587103 on Build Rnor_6.0
UniGene ID
Rn.220523
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130570.1 NP_001124042.1 1 388

View Full Product Details