Gene Transcripts

Gene Symbol
Marco
Entrez Gene ID
367391
Gene Name
macrophage receptor with collagenous structure
Gene Aliases
Chromosome Location
Chr. 16: 85115199 - 85147683 on Build Rnor_6.0
UniGene ID
Rn.83054
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109011.1 NP_001102481.1 16 1606

View Full Product Details