Gene Transcripts

Gene Symbol
USP6
Entrez Gene ID
9098
Gene Name
ubiquitin specific peptidase 6
Gene Aliases
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
Chromosome Location
Chr. 17: 5116438 - 5175032 on Build GRCh38
UniGene ID
Hs.448851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304284.1 NP_001291213.1 31, 32 5058
NM_004505.3 NP_004496.2 23, 24 4527
XM_011524050.1 XP_011522352.1 27, 28 5276
XM_011524051.2 XP_011522353.1 29, 30 4371
XM_011524052.2 XP_011522354.1 28, 29 4485
XM_011524053.2 XP_011522355.1 29, 30 4398
XM_011524054.2 XP_011522356.1 29, 30 4365
XM_011524055.2 XP_011522357.1 27, 28 4292
XM_011524056.2 XP_011522358.1 24, 25 4245
XM_011524057.2 XP_011522359.1 27, 28 4408
XM_011524058.2 XP_011522360.1 28, 29 4474
XM_017025290.1 XP_016880779.1 26, 27 5114
GenBank mRNA AY143550.1 - 23, 24 4527
BX647719.1 - 31, 32 5060
X63546.1 - 22, 23 4412
X63547.1 - 24, 25 4735

View Full Product Details