Gene Transcripts

Gene Symbol
Olr95
Entrez Gene ID
405909
Gene Name
olfactory receptor 95
Gene Aliases
Chromosome Location
Chr. 1: 168478496 - 168479440 on Build Rnor_6.0
UniGene ID
Rn.141892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001024.1 NP_001001024.1 1 772

View Full Product Details