Gene Transcripts
- Gene Symbol
- Znhit3
- Entrez Gene ID
- 497975
- Gene Name
- zinc finger, HIT-type containing 3
- Gene Aliases
- Chromosome Location
- Chr. 10: 72227712 - 72235929 on Build Rnor_6.0
- UniGene ID
- Rn.224655
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_573169.6 |
XP_573169.3
|
4, 5 |
313 |
|
GenBank mRNA |
FQ231514.1 |
- |
4, 5 |
290 |
|
View Full Product Details