Gene Transcripts

Gene Symbol
TBRG4
Entrez Gene ID
9238
Gene Name
transforming growth factor beta regulator 4
Gene Aliases
CPR2, FASTKD4
Chromosome Location
Chr. 7: 45100100 - 45111747 on Build GRCh38
UniGene ID
Hs.231411
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261834.1 NP_001248763.1 2 410
NM_004749.3 NP_004740.2 2 454
NM_030900.3 NP_112162.1 2 454
NM_199122.2 NP_954573.1 2 433
GenBank mRNA AB023165.1 - 2 425
AK001281.1 - 2 402
AK022575.1 - 2 390
AK222943.1 - 2 390
AK300489.1 - 2 369
AL833840.1 - 2 423
BC014918.1 - 2 384 3906670
BC017235.2 - 2 381 3538504
BG703429.1 - 2 419
DA980367.1 - 2 454

View Full Product Details