Gene Transcripts

Gene Symbol
Acer3
Entrez Gene ID
499210
Gene Name
alkaline ceramidase 3
Gene Aliases
Phca, RGD1561254
Chromosome Location
Chr. 1: 163160987 - 163263620 on Build Rnor_6.0
UniGene ID
Rn.19122
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759809.1 XP_008758031.1 2 285
GenBank mRNA FQ230480.1 - 2 238

View Full Product Details