Gene Transcripts

Gene Symbol
NREP
Entrez Gene ID
9315
Gene Name
neuronal regeneration related protein
Gene Aliases
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
Chromosome Location
Chr. 5: 111729303 - 111976931 on Build GRCh38
UniGene ID
Hs.36053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142474.1 NP_001135946.1 3 345
NM_001142475.1 NP_001135947.1 3 375
NM_001142476.1 NP_001135948.1 4 427
NM_001142477.1 NP_001135949.1 4 318
NM_001142478.1 NP_001135950.1 4 294
NM_001142479.1 NP_001135951.1 4 331
NM_001142480.1 NP_001135952.1 4 325
NM_001142481.1 NP_001135953.1 4 234
NM_001142482.1 NP_001135954.1 4 241
NM_001142483.1 NP_001135955.1 2 530
NM_004772.2 NP_004763.1 3 222
XM_006714732.1 XP_006714795.1 2 142
XM_006714733.2 XP_006714796.1 3 123
GenBank mRNA AK298779.1 - 3 270
AK307346.1 - 2 79
AK310585.1 - 2 107
AK315617.1 - 3 196
AL547663.3 - 4 322
BC011050.1 - 4 255 4340005
BC019068.2 - 3 158 4893075
BC072013.1 - 3 181 5922876
BC072443.1 - 3 186 6162335
BX649051.1 - 3 169
DA027062.1 - 3 345
DA078027.1 - 2 255
DA490948.1 - 4 427
DA520221.1 - 4 253
DA526263.1 - 4 415
DA534424.1 - 2 530
DA683910.1 - 4 241
DA694580.1 - 3 240
DA704824.1 - 4 331
DA744980.1 - 4 178
DA752769.1 - 4 234
DA908653.1 - 3 222
DA939543.1 - 4 318
DC365764.1 - 3 375
U30521.1 - 4 253
U36189.1 - 2 66

View Full Product Details