Gene Transcripts

Gene Symbol
Gstt3
Entrez Gene ID
499422
Gene Name
glutathione S-transferase, theta 3
Gene Aliases
RGD1562732
Chromosome Location
Chr. 20: 13817795 - 13825475 on Build Rnor_6.0
UniGene ID
Rn.92581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137643.1 NP_001131115.1 1, 2 575
GenBank mRNA FQ225600.1 - 1, 2 238

View Full Product Details