Gene Transcripts

Gene Symbol
Cyth4
Entrez Gene ID
500906
Gene Name
cytohesin 4
Gene Aliases
Pscd4, RGD1564842
Chromosome Location
Chr. 7: 119820518 - 119845003 on Build Rnor_6.0
UniGene ID
Rn.154985
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130577.1 NP_001124049.1 12 1147
XM_006242031.2 XP_006242093.1 13 1234
XM_006242033.2 XP_006242095.1 12 1151

View Full Product Details