Gene Transcripts

Gene Symbol
Layn
Entrez Gene ID
500996
Gene Name
layilin
Gene Aliases
RGD1564444
Chromosome Location
Chr. 8: 55447203 - 55468565 on Build Rnor_6.0
UniGene ID
Rn.230868
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191997.1 NP_001178926.1 3 190
XM_006243049.2 XP_006243111.1 2 1155
XM_008766328.1 XP_008764550.1 2 1155

View Full Product Details