Gene Transcripts

Gene Symbol
Zmat5
Entrez Gene ID
501926
Gene Name
zinc finger, matrin type 5
Gene Aliases
RGD1559774
Chromosome Location
Chr. 14: 84937843 - 84968898 on Build Rnor_6.0
UniGene ID
Rn.3249
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251427.2 XP_006251489.1 2 1391
XM_577351.5 XP_577351.3 2 382
GenBank mRNA FQ227485.1 - 2 134

View Full Product Details