Gene Transcripts

Gene Symbol
Cst13
Entrez Gene ID
502679
Gene Name
cystatin 13
Gene Aliases
RGD1564873
Chromosome Location
Chr. 3: 143151392 - 143161525 on Build Rnor_6.0
UniGene ID
Rn.158619
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109343.1 NP_001102813.1 3 486
XM_006235162.2 XP_006235224.1 4 1077

View Full Product Details