Gene Transcripts

Gene Symbol
Cdkl4
Entrez Gene ID
503009
Gene Name
cyclin-dependent kinase-like 4
Gene Aliases
Chromosome Location
Chr. 6: 3232278 - 3259066 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764451.1 XP_008762673.1 6 633

View Full Product Details