Gene Transcripts

Gene Symbol
Dram1
Entrez Gene ID
679937
Gene Name
DNA-damage regulated autophagy modulator 1
Gene Aliases
Dram
Chromosome Location
Chr. 7: 28898647 - 28932641 on Build Rnor_6.0
UniGene ID
Rn.1826
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173427.1 NP_001166898.1 6 739
GenBank mRNA BC168648.1 - 6 739
FQ211820.1 - 6 867
FQ223303.1 - 6 799
FQ229264.1 - 6 800

View Full Product Details