Gene Transcripts

Gene Symbol
Sdf2l1
Entrez Gene ID
680945
Gene Name
stromal cell-derived factor 2-like 1
Gene Aliases
Chromosome Location
Chr. 11: 88122271 - 88124513 on Build Rnor_6.0
UniGene ID
Rn.1414
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109433.1 NP_001102903.1 2 420
GenBank mRNA FQ225991.1 - 2 396
FQ230956.1 - 2 397

View Full Product Details