Gene Transcripts

Gene Symbol
Vom2r48
Entrez Gene ID
686145
Gene Name
vomeronasal 2 receptor, 48
Gene Aliases
Chromosome Location
Chr. 4: 156324922 - 156366577 on Build Rnor_6.0
UniGene ID
Rn.217637
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099514.1 NP_001092984.1 3 866

View Full Product Details