Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014705.3 | NP_055520.3 | 10 | 1101 | |
XM_006716188.2 | XP_006716251.1 | 10 | 1088 | ||
XM_006716189.2 | XP_006716252.1 | 10 | 1088 | ||
XM_017012819.1 | XP_016868308.1 | 10 | 1461 | ||
XM_017012820.1 | XP_016868309.1 | 10 | 1024 | ||
XM_017012821.1 | XP_016868310.1 | 10 | 1015 | ||
XM_017012822.1 | XP_016868311.1 | 10 | 1606 | ||
XM_017012823.1 | XP_016868312.1 | 10 | 1015 | ||
XM_017012824.1 | XP_016868313.1 | 10 | 1015 | ||
XM_017012825.1 | XP_016868314.1 | 10 | 1015 | ||
GenBank mRNA | AK309605.1 | - | 10 | 1101 | |
AY233380.1 | - | 10 | 832 | ||
BC117688.2 | - | 10 | 832 | 40072000 | |
BC117689.1 | - | 10 | 832 | 40072006 | |
BC117694.1 | - | 10 | 832 | 40072219 |