Gene Transcripts
- Gene Symbol
- Stath
- Entrez Gene ID
- 689604
- Gene Name
- statherin
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.131908
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY496457.1 |
- |
|
207 |
|
View Full Product Details