Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290474.1 | NP_001277403.1 | 13 | 2242 | |
NM_014807.4 | NP_055622.3 | 13 | 2245 | ||
XM_006718948.3 | XP_006719011.1 | 12 | 2423 | ||
XM_011543076.2 | XP_011541378.1 | 13 | 2741 | ||
XM_017018625.1 | XP_016874114.1 | 12 | 2420 | ||
XM_017018626.1 | XP_016874115.1 | 12 | 2333 | ||
XM_017018627.1 | XP_016874116.1 | 12 | 2330 | ||
XM_017018628.1 | XP_016874117.1 | 13 | 1667 | ||
GenBank mRNA | AB006623.2 | - | 13 | 1707 | |
AB094094.1 | - | 13 | 2213 | ||
AK025223.1 | - | 13 | 2213 | ||
BC003598.1 | - | 5 | 727 | ||
BC010071.2 | - | 7 | 935 | ||
BC021254.2 | - | 7 | 935 | ||
BC022219.1 | - | 13 | 2108 | 3905001 | |
BC033083.1 | - | 7 | 935 | ||
BC052246.1 | - | 5 | 715 | ||
BC075832.1 | - | 7 | 935 | ||
BX537572.1 | - | 7 | 3117 |