Gene Transcripts

Gene Symbol
Ldlrad1
Entrez Gene ID
689088
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
Chromosome Location
Chr. 5: 126812168 - 126821437 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238557.2 XP_006238619.1 7 1239

View Full Product Details