Gene Transcripts

Gene Symbol
Gpr21
Entrez Gene ID
689917
Gene Name
G protein-coupled receptor 21
Gene Aliases
Chromosome Location
Chr. 3: 21764377 - 21767300 on Build Rnor_6.0
UniGene ID
Rn.218692
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109554.1 NP_001103024.1 2 678

View Full Product Details