Gene Transcripts

Gene Symbol
REC8
Entrez Gene ID
9985
Gene Name
REC8 meiotic recombination protein
Gene Aliases
HR21spB, REC8L1, Rec8p
Chromosome Location
Chr. 14: 24171853 - 24180408 on Build GRCh38
UniGene ID
Hs.419259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048205.1 NP_001041670.1 5 967
NM_005132.2 NP_005123.2 6 866
XM_011537421.1 XP_011535723.1 5 1139
XM_017021841.1 XP_016877330.1 5 936
GenBank mRNA AF006264.1 - 5 439
AK001435.1 - 6 866
AK291191.1 - 6 998
AL832973.1 - 5 857
BC004159.2 - 5 523 2822680
BC010887.1 - 5 900 3535425
CR457263.1 - 5 439

View Full Product Details