Gene Transcripts

Gene Symbol
LTB4R
Entrez Gene ID
1241
Gene Name
leukotriene B4 receptor
Gene Aliases
BLT1, BLTR, CMKRL1, GPR16, LTB4R1, LTBR1, P2RY7, P2Y7
Chromosome Location
Chr. 14: 24311499 - 24318036 on Build GRCh38
UniGene ID
Hs.567248
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143919.2 NP_001137391.1 2 439
NM_181657.3 NP_858043.1 2 2047
GenBank mRNA BC004545.2 - 2 450
BT007267.1 - 1 118
D89078.1 - 2 1835
D89079.1 - 2 439
U41070.1 - 2 422
X98356.1 - 2 506

View Full Product Details