Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197181.1 | NP_001184110.1 | 3 | 376 | |
NM_006086.3 | NP_006077.2 | 3 | 319 | ||
GenBank mRNA | AF427491.3 | - | 3 | 196 | |
AK058140.1 | - | 3 | 253 | ||
AK122757.1 | - | 2 | 2278 | ||
AK127418.1 | - | 374 | |||
AK223039.1 | - | 3 | 253 | ||
AK292219.1 | - | 3 | 376 | ||
AK307201.1 | - | 4 | 392 | ||
AK314616.1 | - | 3 | 258 | ||
AU142509.1 | - | 3 | 256 | ||
BC000748.2 | - | 3 | 245 | 2823044 | |
BC001678.2 | - | 2 | 64 | ||
BC003021.1 | - | 3 | 245 | 2823044 | |
CD621827.1 | - | 3 | 273 | ||
DA054466.1 | - | 3 | 319 | ||
S62643.1 | - | 3 | 175 | ||
U47634.1 | - | 3 | 196 |