Gene Transcripts

Gene Symbol
NDUFA11
Entrez Gene ID
126328
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases
B14.7, CI-B14.7
Chromosome Location
Chr. 19: 5891276 - 5904013 on Build GRCh38
UniGene ID
Hs.406062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193375.1 NP_001180304.1 2 471
NM_175614.4 NP_783313.1 2 471
NR_034166.2 3 591
GenBank mRNA AJ539081.1 - 2 166
AK127692.1 - 2 244
AL546175.3 - 3 332
AY726561.1 - 2 227
BC069045.1 - 2 226 6503302
BQ054257.1 - 2 227
BQ671956.1 - 2 245
BQ955016.1 - 2 217

View Full Product Details