Gene Transcripts

Gene Symbol
CLEC18A
Entrez Gene ID
348174
Gene Name
C-type lectin domain family 18 member A
Gene Aliases
MRCL, MRCL1, MRLP2
Chromosome Location
Chr. 16: 69950233 - 69968478 on Build GRCh38
UniGene ID
Hs.592064
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136214.2 NP_001129686.1 11, 12 1349
NM_001271197.1 NP_001258126.1 11, 12 1429
NM_182619.3 NP_872425.2 11, 12 1455
XM_005255934.1 XP_005255991.1 10, 11 1398
XM_017023206.1 XP_016878695.1 10, 11 1398
XM_017023207.1 XP_016878696.1 9, 10 1228
XM_017023208.1 XP_016878697.1 10, 11 1581
GenBank mRNA AF428259.1 - 10, 11 1466
AF521893.1 - 11, 12 1468
AK074773.1 - 10, 11 1283
AK289884.1 - 10, 11 1397
BC078143.1 - 11, 12 1331 5761001
BC141808.1 - 11, 12 1429 40147524

Gene Symbol
CLEC18B
Entrez Gene ID
497190
Gene Name
C-type lectin domain family 18 member B
Gene Aliases
MRCL2
Chromosome Location
Chr. 16: 74408631 - 74422248 on Build GRCh38
UniGene ID
Hs.454670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005255976.2 XP_005256033.1 10, 11 1488
XM_011523093.2 XP_011521395.1 9, 10 1194
XM_017023255.1 XP_016878744.1 11, 12 1665
GenBank mRNA BM695392.1 - 6, 7 622

Gene Symbol
CLEC18C
Entrez Gene ID
283971
Gene Name
C-type lectin domain family 18 member C
Gene Aliases
MRCL, MRCL3
Chromosome Location
Chr. 16: 70173322 - 70187147 on Build GRCh38
UniGene ID
Hs.556045
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173619.3 NP_775890.2 10, 11 1317
XM_006721179.1 XP_006721242.1 10, 11 1413
XM_011523034.1 XP_011521336.1 11, 12 1659
XM_011523035.1 XP_011521337.1 10, 11 1387
XM_011523036.2 XP_011521338.1 9, 10 1243
GenBank mRNA AF448856.1 - 10, 11 1465
AL833339.1 - 7, 8 2687
BC039068.1 - 8, 9 2371 5188643

View Full Product Details