Gene Transcripts

Gene Symbol
NIPAL4
Entrez Gene ID
348938
Gene Name
NIPA like domain containing 4
Gene Aliases
ARCI6, ICHTHYIN, ICHYN
Chromosome Location
Chr. 5: 157460019 - 157474722 on Build GRCh38
UniGene ID
Hs.4285
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099287.1 NP_001092757.1 2 466
NM_001172292.1 NP_001165763.1 2 466
XM_011534552.1 XP_011532854.1 2 378
GenBank mRNA AK026158.1 - 2 270
AK296972.1 - 2 466
AK308830.1 - 2 270
AK309242.1 - 2 378
DB153698.1 - 2 417

View Full Product Details