Gene Transcripts

Gene Symbol
RMND5B
Entrez Gene ID
64777
Gene Name
required for meiotic nuclear division 5 homolog B
Gene Aliases
GID2, GID2B
Chromosome Location
Chr. 5: 178130532 - 178148570 on Build GRCh38
UniGene ID
Hs.27222
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005265971.4 XP_005266028.1 1 98
XM_017009727.1 XP_016865216.1 1 98
XM_017009728.1 XP_016865217.1 1 98

View Full Product Details