Gene Transcripts

Gene Symbol
C16orf90
Entrez Gene ID
646174
Gene Name
chromosome 16 open reading frame 90
Gene Aliases
Chromosome Location
Chr. 16: 3493484 - 3495521 on Build GRCh38
UniGene ID
Hs.632224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080524.1 NP_001073993.1 3 777
XM_005255502.3 XP_005255559.1 3 843

View Full Product Details