Gene Transcripts

Gene Symbol
CLEC3A
Entrez Gene ID
10143
Gene Name
C-type lectin domain family 3 member A
Gene Aliases
CLECSF1
Chromosome Location
Chr. 16: 78022546 - 78032106 on Build GRCh38
UniGene ID
Hs.177936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005752.4 NP_005743.4 2 209
GenBank mRNA AK313316.1 - 2 209
AY358376.1 - 2 161
BC104414.2 - 2 198 40003144
BC104415.2 - 2 198 40003149
DA998903.1 - 2 209

View Full Product Details