Gene Transcripts

Gene Symbol
SLC35B1
Entrez Gene ID
10237
Gene Name
solute carrier family 35 member B1
Gene Aliases
UGTREL1
Chromosome Location
Chr. 17: 49700934 - 49708173 on Build GRCh38
UniGene ID
Hs.154073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278784.1 NP_001265713.1 3 411
NM_005827.2 NP_005818.2 3 436
XM_005256929.3 XP_005256986.1 4 351
XM_006721632.3 XP_006721695.1 4 555
XM_011524179.2 XP_011522481.1 2 321
XM_011524180.2 XP_011522482.1 3 287
GenBank mRNA AK098056.1 - 3 618
AK293563.1 - 3 436
AK296073.1 - 3 405
BC011888.2 - 3 415 3832062
BX641166.1 - 4 383
CX164572.1 - 1 43
D87989.1 - 3 405

View Full Product Details