Gene Transcripts

Gene Symbol
PCDHGC3
Entrez Gene ID
5098
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
PC43, PCDH-GAMMA-C3, PCDH2
Chromosome Location
Chr. 5: 141476002 - 141512977 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002588.3 NP_002579.2 1 538
NM_032402.1 NP_115778.1 1 538
GenBank mRNA AF152337.1 - 1 423
AF152524.1 - 1 423
BC019299.1 - 1 537 2905509
BC026218.1 - 1 527 4153558
L11373.1 - 1 537

View Full Product Details