Gene Transcripts

Gene Symbol
PI4KAP2
Entrez Gene ID
375133
Gene Name
phosphatidylinositol 4-kinase alpha pseudogene 2
Gene Aliases
Chromosome Location
Chr. 22: 21472998 - 21517491 on Build GRCh38
UniGene ID
Hs.448225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003700.1 6 766
GenBank mRNA AK126860.1 - 4 2730
AK298744.1 - 6 787
AK299734.1 - 6 631
BC020225.1 - 6 764
BC139841.1 - 4 487
BX407261.2 - 1 48
CR456378.1 - 4 505
CX751888.1 - 1 71

Gene Symbol
PI4KA
Entrez Gene ID
5297
Gene Name
phosphatidylinositol 4-kinase alpha
Gene Aliases
PI4K-ALPHA, PIK4CA, PMGYCHA, pi4K230
Chromosome Location
Chr. 22: 20707691 - 20858812 on Build GRCh38
UniGene ID
Hs.529438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_058004.3 NP_477352.3 36 4327
XM_005261634.1 XP_005261691.1 35 4234
XM_005261635.1 XP_005261692.1 36 4327
XM_017028829.1 XP_016884318.1 35 4261
XM_017028830.1 XP_016884319.1 22 2483
GenBank mRNA AB210002.1 - 36 4297
AF012872.1 - 35 4066
AK302424.1 - 36 4255
AK314781.1 - 8 981
BC018120.1 - 6 644
BC053654.1 - 5 620
L36151.1 - 4 613

Gene Symbol
PI4KAP1
Entrez Gene ID
728233
Gene Name
phosphatidylinositol 4-kinase alpha pseudogene 1
Gene Aliases
Chromosome Location
Chr. 22: 18533864 - 18548828 on Build GRCh38
UniGene ID
Hs.592456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003563.1 4 499
GenBank mRNA AK096724.1 - 6 725
AK292695.1 - 5 590
BC072398.1 - 6 775
BC131538.1 - 4 499

View Full Product Details