Gene Transcripts

Gene Symbol
Rhox9
Entrez Gene ID
104384
Gene Name
reproductive homeobox 9
Gene Aliases
1600026O01Rik, Gpbox, Psx2
Chromosome Location
Chr. X: 37899097 - 37901770 on Build GRCm38
UniGene ID
Mm.156958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023894.1 NP_076383.1 2 533
GenBank mRNA AF201698.1 - 2 533
BC099451.1 - 2 472 30716051
BC099963.1 - 2 467 30083999
DQ058646.1 - 2 533

View Full Product Details