Gene Transcripts

Gene Symbol
Kazald1
Entrez Gene ID
107250
Gene Name
Kazal-type serine peptidase inhibitor domain 1
Gene Aliases
AI842353, Bono1, IGFBP-rP10
Chromosome Location
Chr. 19: 45074485 - 45079289 on Build GRCm38
UniGene ID
Mm.24417
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178929.4 NP_849260.2 5 1254
GenBank mRNA AK030678.1 - 5 1253
AK155095.1 - 6 1499

View Full Product Details