Gene Transcripts

Gene Symbol
Grk4
Entrez Gene ID
14772
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
A830025H08Rik, GRK, Gprk2l, Gprk4
Chromosome Location
Chr. 5: 34660287 - 34755305 on Build GRCm38
UniGene ID
Mm.117076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080743.1 NP_001074212.1 2, 3 716
NM_019497.2 NP_062370.2 2, 3 716
GenBank mRNA AF040745.1 - 2, 3 142
AK020792.1 - 2, 3 716
AK078475.1 - 2, 3 588
AK132685.1 - 2, 3 717
BC150691.1 - 2, 3 240 9087602
BY719486.1 - 2, 3 588

View Full Product Details