Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133209.2 | NP_573472.2 | 2 | 534 | |
GenBank mRNA | AB122024.2 | - | 2 | 429 | |
AJ400847.1 | - | 2 | 483 | ||
BC145880.1 | - | 2 | 472 | 40131183 | |
BC145906.1 | - | 2 | 472 | 40131209 | |
BC172053.1 | - | 2 | 472 | 9054339 |