Gene Transcripts

Gene Symbol
Stfa1
Entrez Gene ID
20861
Gene Name
stefin A1
Gene Aliases
Stefin-3, Stf-3, Stf1, Stf3, Stfa3
Chromosome Location
Chr. 16: 36277148 - 36285371 on Build GRCm38
UniGene ID
Mm.327618
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082543.1 NP_001076012.1 2 196
GenBank mRNA BY113926.1 - 2 196
M92417.1 - 2 165

View Full Product Details